Learn More
Viro Labs ADHP SOLUTION, SUPER RED 80ML
Supplier:  Viro Labs TRED80
Encompass
SuperRed, one component, ready-to-use ADHP solution for fluorescence ELISA with HRP conjugates 80ml
Catalog No. NC2109323