Learn More
Medchemexpress LLC Adipic acid-d8 | 52089-65-3 | 99.7% | 154.19 g/mol | C6H2D8O4 | 10 MG

Supplier: Medchemexpress LLC HYW017522S410MG
Adipic acid-d8 is the deuterium-labeled isotopologue of adipic acid (hexanedioic acid) used as an internal standard and reference material for analytical chemistry and mass spectrometry. It provides a predictable mass shift that supports accurate quantitation, method validation, and trace-level analysis in LC and GC workflows.
- Deuterium-labeled (d8) adipic acid for mass-shifted internal standardization.
- CAS number 52089-65-3 for unambiguous identification.
- Chemical formula C6H2D8O4 and molecular weight 154.19 g/mol.
- High chemical purity suitable for analytical use (reported 99.65%).
- Compatible with LC-MS/MS and GC-MS workflows.
- Supplied in small pack sizes appropriate for method development and standards preparation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.