Learn More
Medchemexpress LLC Adipic acid-d8 (hexanedioic acid-d8) | 52089-65-3 | 99.7% | 154.19 | C6H2D8O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW017522S450MG
Encompass_Preferred
Adipic acid-d8 (hexanedioic acid-d8) is a deuterium-labeled adipic acid supplied as a 50 mg research standard for analytical and mechanistic studies. It is intended for use as a tracer and internal standard in quantitative techniques such as NMR, GC-MS, and LC-MS, and is provided at high chemical purity for accurate quantitation.
  • High isotopic labeling for accurate quantitation.
  • Suitable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (99.65%) for analytical applications.
  • Compact 50 mg pack for research-scale use.
  • Molecular weight 154.19 and formula C6H2D8O4.
Catalog No. 50-003-01787