Learn More
CPC Scientific Pyr-Leu-Thr-Phe-Thr-Ser-Ser-Trp-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific INSE001A
Encompass
AKH, a peptide hormone which stimulates synthesis of 1,2-diacyl-sn-glycerols in the Manduca sexta fat body.ONE-LETTER SEQUENCE: Pyr-LTFTSSWG-NH2MOLECULAR FORMULA: C47H65N11O14MOLECULAR WEIGHT:1008.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [99886-31-4], RESEARCH AREA: ImmunomodulationREFERENCES: R.Ziegler et al., Biochem. Biophys. Res. Commun., 133, 337 (1985); H.Jaffe et al., Biochem. Biophys. Res. Commun., 135, 622 (1986); E.L.Arrese and M.A.Wells, J. Lipid Res., 38, 68 (1997)
Catalog No. 50-210-9602