Learn More
Medchemexpress LLC HY-15848 10mg Medchemexpress, AdipoRon CAS:924416-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1584810MG
Encompass_Preferred
Medchemexpress, HY-15848 10mg AdipoRon CAS:924416-43-3 AdipoRon is an orally active adiponectin receptor (AdipoR) agonist, binding to AdipoR1 and AdipoR2 with Kds of 1.8 and 3.1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9958