Learn More
Abcam AdipoRon, Adiponectin receptor agonist, 10MG
Supplier:  Abcam AB14486710MG
Encompass_Preferred
MW 428.5 Da, Purity >98%. Adiponectin receptor agonist (KD values are 1.8 and 3.1 μM for AdipoR1 and AdipoR2, respectively). AMPK and PPARα signaling activator. Shows antiobesogenic, antidiabetic and cardioprotective effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5464