Learn More
Cayman Chemical Adipose Triglyceride Lipase P
SDP
Supplier:  Cayman Chemical 100064091 EA
Encompass_Preferred
Immunogen: Synthetic peptide from an internal region of human ATGL • Host: Rabbit • Species Reactivity: (+) Human, mouse, and rat • Applications: IHC (paraffin-embedded tissue) and WB
Catalog No. 50-288-7020