Learn More
Cayman Chemical AdIpostatIn A 5mg
SDP
Supplier:  Cayman Chemical 221635 MG
Encompass_Preferred
A GPDH inhibitor (IC50 = 4.1 µM); prevents triglyceride accumulation in 3T3-L1 cells when applied at 5-7.5 µM; cytotoxic against fibroblast carcinoma KB cell lines (IC50 = 10.6 µM)
Catalog No. 50-298-5707