Learn More
Teledyne ISCO, Inc ADJ TORRNT SOLID LOAD CAP 750G
Supplier:  Teledyne ISCO, Inc 605247009
Encompass
Adjustable Solid Load Cartridge Cap (SLCC), fits 375 and 750 gram sample load cartridges. Includes loading rod. For use on CombiFlash Torrent only.
Catalog No. 50-201-557