Learn More
Medchemexpress LLC ADL-5859 hydrochloride | 850173-95-4 | 99.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1304410MM/1ML
Encompass_Preferred
ADL-5859 hydrochloride (compound 20) is a selective and orally active δ opioid receptor (DOR) agonist with a Ki of 0.84 nM and an EC50 value of 20 nM. It also exhibits inhibitory activity to the hERG channel with an IC50 value of 78 μM. ADL-5859 hydrochloride can be utilized for pain research.
  • Selective and orally active δ opioid receptor agonist
  • Ki of 0.84 nM
  • EC50 value of 20 nM
  • Inhibitory activity to the hERG channel with an IC50 value of 78 μM
  • Useful for pain research
Catalog No. 50-003-40342