Learn More
Medchemexpress LLC Adomeglivant (LY2409021) | 1488363-78-5 | MFCD30343860 | 99.6% | 555.63 g/mol | C32H36F3NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1990410MG
Encompass_Preferred
Adomeglivant is a small-molecule, orally active allosteric antagonist of the glucagon receptor used in research on glucose regulation and type 2 diabetes. It is applied as an investigational tool to study glucagon signaling and metabolic effects in preclinical and clinical settings.
  • Potent, selective glucagon receptor allosteric antagonist.
  • High purity suitable for analytical and research use.
  • Solid white to off-white powder, easy to handle.
  • Highly soluble in DMSO for solution preparation.
  • Stable under recommended storage conditions for long-term use.
  • Used in preclinical and clinical research on glucose metabolism.
Catalog No. 50-202-9301