Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp ADP [Adenosine-5'-diphosphate, disodium salt dihydrate], 1 Gram
SDP

Supplier:  Research Products International Corp A112201.0

Encompass_Preferred

CAS Number: 16178-48-6
Molecular Formula: C10H13N5O10P2Na2 • 2H2O
Molecular Weight: 507.2

Catalog No. 50-136-7996


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.