Learn More
Apexbio Technology LLC Adenosine-5'-diphosphate(Synonyms: ADP, Adenosine diphosphate, 5'-ADP, Adenosine 5'-pyrophosphate), 250mg, CAS: 58-64-0.
SDP
Supplier:  Apexbio Technology LLC C4341250
Encompass_Preferred
Adenosine-5 -diphosphate (CAS 58-64-0) commonly known as ADP is an endogenous nucleotide serving as an agonist of purinergic receptors It activates various P2 receptor subtypes exhibiting an IC50 of 67 nM at P2X2/3 receptors ADP results from ATP hydrolysis mediated by ATPases and can be reconverted to ATP via ATP synthases or further processed to AMP and other metabolites Through receptor modulation ADP influences cellular functions including neural stem cell proliferation vascular response apoptosis cytokine secretion and kinase-driven phosphorylation pathways thus playing a critical role in purinergic signaling research
Catalog No. 50-000-61152