Learn More
Eurofins Discoverx ADP Hunter™ Plus 8ml (800 dp, 384-well)
Supplier:  Eurofins Discoverx 900083
Encompass
The ADP Hunter™ Plus kit is a detection kit for assaying kinase activity via production of ADP. This detection kit contains reagents for 2 plates (384-well).
Catalog No. NC2824284