Learn More
Medchemexpress LLC Adpm06 | 490035-90-0 | 95.0% | 715.19 g·mol⁻¹ | C34H24BBr2F2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135475MG
Encompass_Preferred
ADPM06 is an azadipyrromethene research compound developed as a non-porphyrin photodynamic therapy (PDT) sensitizer. It exhibits micromolar potency in tumor cell lines and induces apoptosis, and is supplied for laboratory research and preclinical investigation.
  • Non-porphyrin photodynamic sensitizer for PDT research.
  • Demonstrates micromolar IC50 potency and induces apoptosis in tumor cells.
  • Molecular formula C34H24BBr2F2N3O2; molecular weight 715.19 g·mol⁻¹.
  • High purity (≈95%) suitable for research applications.
  • Solid, brown to black appearance; stable when stored under recommended conditions.
  • Available in small research quantities with variable lead times.
Catalog No. 50-002-22009