Learn More
Eurofins Discoverx ChemiScreen α1D Adrenergic Receptor Stable Cell Line, 2 vials. (HTS216C)
Supplier:  Eurofins Discoverx HTS216C
Encompass
The ChemiScreen α1D Adrenergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1D GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1D (lacking residues 2-79) alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of ADRA1D, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6065
Only null left
Add to Cart
Request a Quote