Learn More
Medchemexpress LLC Adrenalone (hydrochloride) | 62-13-5 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1308100MG
Encompass_Preferred
Adrenalone hydrochloride is an adrenergic agonist used as a topical vasoconstrictor and hemostatic. It is also an inhibitor of dopamine β oxidase and is chemically similar to known norepinephrine transporter (NET) ligands with an IC50 of 36.9 μM. It is a topical nasal decongestant, hemostatic, and vasoconstrictor, and a keton form of the natural substrate epinephrine. Adrenalone hydrochloride inhibits the activity of dopamine β-oxidase and substrate transport by NET.
  • Adrenergic agonist
  • Topical vasoconstrictor and hemostatic
  • Inhibitor of dopamine β oxidase
  • Chemically similar to norepinephrine transporter (NET) ligands (IC50 of 36.9 μM)
  • Topical nasal decongestant
  • Keton form of natural substrate epinephrine
Catalog No. 50-004-11842