Learn More
Medchemexpress LLC Adrenocorticotropic Hormone (1-13) | 22006-64-0 | 99.6% | 1623.83 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP15555MG
Encompass_Preferred
ACTH (1-13) is a 13-aa peptide with cytoprotective effects in the model of ethanol-induced gastric lesions in rats. It has also shown immunomodulatory effects. Adrenocorticotropic hormone (ACTH) is a tropic hormone produced by the anterior pituitary that regulates cortisol and androgen production and is associated with Addison disease, Cushing syndrome, and Cushing disease.
  • Cytoprotective effects in ethanol-induced gastric lesions
  • Immunomodulatory effects
  • Regulates cortisol and androgen production
  • Associated with Addison disease, Cushing syndrome, and Cushing disease
Catalog No. 50-004-23531