Learn More
Genscript Corporation Adrenomedullin (AM) (1-52), human
Supplier:  Genscript Corporation RP112880.5
Encompass
Adrenomedullin (1-52) is a potent hypotensive peptide hormone with some structural similarity to calcitonin gene-related peptide. Adrenomedullin (1-52) has vasodilatory properties.
Catalog No. 50-275-073