Learn More
Medchemexpress LLC Adrogolide hydrochloride | 166591-11-3 | 99.6% | 435.96 g/mol | C22H26ClNO4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11895650MG
Encompass_Preferred
Adrogolide hydrochloride is a chemically stable prodrug that is converted to the dopamine D1 receptor agonist A-86929 and is used in neuropharmacology research, including Parkinson's disease and cognition models.
  • Chemically stable prodrug that converts to dopamine D1 agonist A-86929.
  • Used in neuropharmacology and Parkinson's disease research models.
  • High purity (99.6%) with enantiomeric excess reported as 100%.
  • Soluble in DMSO (approximately 66.7 mg/mL with sonication and warming).
  • Offered in research-scale pack sizes, including a 50 MG pack.
Catalog No. 50-002-29026