Learn More
Medchemexpress LLC HY-12885B 10mg , ADU-S100 (ammonium salt) CAS:1638750-96-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12885B10MG
Encompass_Preferred
HY-12885B 10mg ADU-S100 (ammonium salt) CAS: 1638750-96-5 ADU-S100 ammonium salt (MIW815 ammonium salt), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0670