Learn More
Chemie Tek ADU-S100 (MIW815), 1638750-95-4, MFCD32215340, 2mg
SDP
Supplier:  Chemie Tek ADUS1002MG
Encompass_Preferred
Mol. Formula: C20H22N10O10P2S2∙2Na, 99.9% Pure, MW: 734.5, in vivo Grade Material, ADU-S100 (MIW815) is a synthetic cyclic dinucleotide (CDN) agonist (activator) of Stimulator of Interferon Genes (STING), a receptor crucial to activate the innate (endogenous) immune system. ADU-S100 (MIW815) activates all known human and mouse STINGs, and effectively induces the expression of cytokines and chemokines, leading to a robust and durable antigen-specific T-cell mediated immune response against cancer cells.
Catalog No. NC1690588