Learn More
Cellutron Adumyt kit
SDP
Supplier:  Cellutron AC7018
Encompass_Preferred
With this simplified Adumyt Isolation System (which does not requires the Langendorf perfusion apparatus), scientists will find it much more easier to obtain high quality, Ca2+ tolerated cardiomyocytes for their important experiments. This system eliminates the technical difficulties with perfusion and cannulation of small hearts, especially from mouse and other small animals.
Catalog No. NC1829815