Learn More
Analytical Sales & Services Adv. 96RW Pattern Sealing Films, PTFE Material, -75ºC to +125ºC Seal Range, 100 Films/Pack
Supplier:  Analytical Sales & Services 96823
Encompass
The Analytical Sales Advantage 96 Round Well Pattern Sealing Films offer superior sealing solutions for laboratory applications. These black adhesive PTFE films ensure a firm and chemical-resistant seal across various substrates, protecting samples from contamination and evaporation. With outstanding temperature and solvent resistance, these films excel in demanding protocols like PCR and are easy to pierce for autosamplers, differentiating them with their adhesive-free target areas.
Catalog No. NC0679879