Learn More
Analytical Sales & Services ADV PREASSEMBLED VIAL KIT 72PK
Supplier:  Analytical Sales & Services ADV376720
Encompass
Advantage Pre-Assembled Vial Kit Contains: 3000 Class 20mL Clear Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners
Catalog No. NC1956595