Learn More
Biomatik Corporation Advanced Glycation End Product (Age) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29357
Encompass_Preferred
Advanced Glycation End Product (Age) Monoclonal Antibody is a Universal Monoclonal Antibody to Advanced Glycation End Product (AGE). The Advanced Glycation End Product (Age) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Universal. Applications: WB, IHC, ICC, IP. Target Synonyms: Advanced Glycation End Product (AGE). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7570