Learn More
Takara Bio Advantage® 2 Polymerase Mix, 100 Rxns
SDP
Supplier:  Takara Bio 639201
Encompass
Advantage 2 Polymerase Mix is a hot-start PCR polymerase mix that contains Takara's Titanium Taq DNA Polymerase (with TaqStart Antibody), and a minor amount of a proofreading polymerase for a balance of higher yield and higher fidelity than wild-type Taq. The Mix is ideal for amplification of long templates up to 18 kb and complex genomic DNA up to 6 kb. Two different optimized buffers are supplied with the polymerase mix. dNTPs need to be purchased separately. Enough enzyme mix and PCR buffer are supplied for 100 PCR reactions of 50 μl each.
Catalog No. NC9557658