Learn More
Takara Bio Advantage® 2 PCR Kit, 100 Rxns
SDP
Supplier:  Takara Bio 639206
Encompass
This kit is based on Takara's Advantage 2 Polymerase Mix. It contains all reagents needed for a hot-start PCR, including control template and primer mix. This kit is ideal for PCR applications that require high fidelity, such as cDNA amplification or library construction. Enough reagents are supplied for 100 PCR reactions of 50 μl each. 100 Rxns
Catalog No. NC9566392