Learn More
Cole-Parmer Advantec PF05055MM PTFE Depth Filter, 5 m pore, 55-mm diameter; pack of 20
Supplier:  Cole-Parmer 3624026
Encompass
Pure PTFE fibers are sintered to improve handling characteristics and minimize fiber slough-off and downstream contamination. Recommended for filtering hot acids, strong solvents, and for applications where media contamination must be at a critically low level. Use these hydrophobic filters to separate aqueous and non-aqueous phases of mixtures as well as for air/gas filtration or venting. Temperature range is –120 to 260C (–184 to 436F).
Catalog No. NC0079216