Learn More
Analytical Sales & Services 96-Well Photoredox Block, Anodized Aluminum, Reduces Solvent Loss, 1 mL
Supplier:  Analytical Sales & Services 96973
Encompass
The Analytical Sales & Services 96-Well Photoredox Block Assembly is designed for high-throughput screening in chemistry. Made from anodized aluminum, it offers enhanced reproducibility and efficiency in experiments while minimizing solvent loss even at high temperatures. This specialized block enables parallel synthesis and is compatible with standard glass shell vials, making it an essential tool for demanding experimental conditions. Ideal for researchers looking to optimize their workflows in photoredox reaction setups.
Catalog No. NC0530509