Learn More
Aviva Systems Biology Corp AEBP1 Antibody - N-terminal region (ARP31592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AEBP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human AEBP1. Alternate names: ACLPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54275