Learn More
Research Products International Corp AEBSF [4-(2-Aminoethyl)-benzenesulfonyl fluoride, HCl], 100 Milligrams
SDP
Supplier:  Research Products International Corp A200100.1
Encompass_Preferred
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.
Catalog No. 50-213-114