Learn More
AG Scientific Inc AEBSF HCl, 250 MG
Supplier:  AG Scientific Inc A1018250MG
Encompass_Preferred
AEBSF HCl (also called Pefabloc) is a broad spectrum, water soluble, irreversible serine protease inhibitor. Inhibition constants are similar to those of PMSF and DFP. AEBSF has been shown to inhibit trypsin, chymotrypsin, plasmin kallikrein and thrombin. As an alternative to PMSF and DFP, AEBSF offers lower toxicity, improved solubility in water and improved stability in aqueous solutions. It has been shown to inhibit the constitutive production of beta amyloid (A beta) in five different human cell lines, both neural and non-neural.AEBSF also inhibits activation of TGFβ (transforming growth factor beta), moderately inhibits dipeptidyl peptidase IV (DPP IV) and activates JNK and p38 MAP kinases.CAS Number: 30827-99-7Molecular Weight: 239.5Chemical Formula: C8H11ClFNO2SSolubility: Clear colorless solution in H2O 50mg/mlStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7007