Learn More
Medchemexpress LLC HY-12821 100mg Medchemexpress, AEBSF (hydrochloride) CAS:30827-99-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12821100MG
Encompass_Preferred
Medchemexpress, HY-12821 100mg AEBSF (hydrochloride) CAS:30827-99-7 AEBSF hydrochloride is an irreversible inhibitor of serine proteases, such as chymotrypsin, kallikrein, plasmin, thrombin, and trypsin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1158