Learn More
Abcam AEBSF hydrochloride, serine protease inhibitor, 200MG
Supplier:  Abcam AB141403200MG
Encompass_Preferred
MW 239.7 Da, Purity >97%. Broad spectrum, irreversible serine protease inhibitor. Inhibits proteases like chymotrypsin, kallikrein, plasmin, thrombin and trypsin. Stable, non-toxic alternative to PMSF (ab141302).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6647