Learn More
Medchemexpress LLC Aeide-C1-NHS ester | 824426-32-6 | 98.0% | C6H6N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140755
Encompass_Preferred
Aeide-C1-NHS ester is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Alkyl/ether-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes CuAAc reaction
  • Undergoes SPAAC reactions
Catalog No. 50-003-52683