Learn More
ABclonal Technology AER61 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2116620UL
Encompass_Preferred
This gene encodes an enzyme that acts in the lumen of the endoplasmic reticulum to catalyze the transfer of N-acetylglucosamine to serine or threonine residues of extracellular-targeted proteins This enzyme modifies proteins containing eukaryotic growth factor (EGF)-like domains including the Notch receptor thereby regulating developmental signalling Mutations in this gene have been observed in individuals with Adams-Oliver syndrome 4 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07576