Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC AERO DUSTER (ORDER MIN 4)AE
SDP
Supplier:  MILLER STEPHENSON CHEMICAL COMPANY INC MS222NEA
Encompass_Preferred
AERO DUSTER,14 oz,Nonflamable,The convienient way to remove microscopic dust.Recommended for electromechanical instruments,computer equipment,rotating instruments and optics.High purity.
Catalog No. NC9698129