Learn More
ATCC Aeromonas salmonicida; subsp. salmonicida
Supplier:  ATCC 33658
Encompass
Aeromonas salmonicida subsp. salmonicida strain NCMB 1102 [19291, 20] is a whole-genome sequenced type strain that was isolated from salmon (Salmo salar). This bacterial culture can be used in susceptibility disc testing and as a quality control strain.
Catalog No. 50-238-1864