Learn More
TARGETMOL CHEMICALS INC AES-135 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T102555MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.AES-135 is a potent HDAC inhibitor, inhibits HDAC3, HDAC6, HDAC11 (IC50s 654, 190, and 636 nM) with anti-tumor activity. Purity 98.01%
Catalog No. 50-288-0623