Learn More
Selleck Chemical LLC AF-353 S0405-1G
SDP
Supplier:  Selleck Chemical LLC S04051G
Encompass_Preferred
AF-353 (Ro-4) is a potent selective and orally bioavailable antagonist of P2X3 receptor and P2X2/3 receptor with pIC50 of 8 06 8 05 and 7 41 for human P2X3 rat P2X3 and human P2X2/3 respectively
Catalog No. 50-000-41116