Learn More
TARGETMOL CHEMICALS INC AF38469 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ00861MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AF38469 is a new, specific and orally bioavailable Sortilin inhibitor (IC50 330 nM). Purity 99.18%
Catalog No. 50-288-0330