Learn More
Biolegend Alexa Fluor 647 anti-human CD54
Supplier:  Biolegend 353114
Encompass_Preferred
Alexa Fluor 647 anti-human CD54 [HA58]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1689815