Learn More
Biolegend Alexa Fluor 647 Goat anti-hamster (Armenian) IgG
Supplier:  Biolegend 405510
Encompass_Preferred
Alexa Fluor 647 Goat anti-hamster (Armenian) IgG [Poly4055]; Isotype: Goat Polyclonal IgG; Reactivity: Armenian Hamster; Apps: FC, ICFC; Size: 100 ug
Catalog No. NC1302926