Learn More
Medchemexpress LLC AF647-NHS ester | 407627-60-5 | 99.7% | 956.09 | C40H49N3O16S4 | 10 MG

Supplier: Medchemexpress LLC HYD111910MG
AF647-NHS ester is an N-hydroxysuccinimide ester of a far-red fluorescent dye used to covalently label amine-containing biomolecules for fluorescence imaging and detection. It is supplied as a solid optimized for conjugation to proteins, antibodies, and peptides.
- Far-red excitation at 635 nm (instantaneous 620 nm) suitable for 633-647 nm lasers.
- High purity by HPLC: 99.74%.
- Molecular weight 956.09; formula C40H49N3O16S4.
- Appearance: dark purple to black solid.
- Storage: long-term at 4 °C sealed; in solvent, -80 °C up to 6 months.
- Supplied as 10 MG vials.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.