Learn More
Medchemexpress LLC AF647-NHS ester triTEA | 407627-61-6 | 99.7% | 1259.66 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD1119C50MG
Encompass_Preferred
AF647-NHS ester is an analogue of Alexa Fluor 647 (AF647). It features an excitation wavelength (λex) of 635 nm for conventional fluorescence detection and 620 nm for instantaneous detection. This compound's NHS ester group enables covalent binding to molecules containing amino groups, such as proteins and antibodies.
  • Fluorescent properties suitable for various detection methods.
  • Reactive NHS ester group for covalent attachment to amino groups.
  • Enables labeling of proteins, antibodies, and other biomolecules.
  • Excitation at 635 nm (conventional) and 620 nm (instantaneous).
  • Available in multiple quantities for research needs.
Catalog No. 50-004-31342