Learn More
Biolegend Alexa Fluor 700 anti-mouse CD107a (LAMP-1)
Supplier:  Biolegend 121628
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1675932