Learn More
Enzo Life Sciences Aflatoxin B1 (5mg). CAS: 1162-65-8
SDP
Supplier:  Enzo Life Sciences ALX630093M005
Encompass_Preferred
Naturally occuring mycotoxin produced by many species of Aspergillus. Metabolized by the liver to the reactive intermediate aflatoxin M1. Exposure may produce acute necrosis, cirrhosis and carcinoma of the liver. Induces DNA damage. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO (20mg/ml) or methylene chloride (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9620