Learn More
Cayman Chemical AflatoxIn M1 100ug
SDP
Supplier:  Cayman Chemical 11297100 UG
Encompass_Preferred
An active metabolite of aflatoxin B1; induces hepatocyte hyperplasia and hepatocellular carcinoma tumor formation in rats at 1 mg/kg; increases hepatic total lipid levels and inhibits hepatic mitochondrial and cytoplasmic protein synthesis in rats
Catalog No. 50-295-2280