Learn More
New England Biolabs, Inc. AflIII – 1250 units
SDP
Supplier:  New England Biolabs, Inc. R0541L
Encompass
A restriction endonuclease that recognizes the sequence A^CRYG_T.
Catalog No. 50-812-212